Advantec Nobuto Filter Strips
Ideal for collection applications
  • Dried samples can be analyzed, stored, or mailed without refrigeration
Collect blood on the narrow end of the strip and place on the drying rack for easy and efficient sample handling. Each strip can hold approximately 0.1 mL of blood or 0.04 mL of serum. Strips are produced with high-purity cellulose paper with a fine, textile like structure—ideal for uniform sampling. Typical blood collection applications include toxoplasmosis (HA), hanta virus, new castles disease (HI), canine distemper (VN), measles (Rubeola), Japanese encephalitis (HI, VN), mycoplasmosis (AG), and hog cholera (VN).
5,000.00 - ₹24,500.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Advantec 800700 Nobuto Blood Filter Strip, 100/pk 0664440 Mfr # 49010010
COMPARE
Nobuto Blood Filter Strip, 100/pk
₹5,000.00INR / Pkg of 100
Advantec 800701 Nobuto Filter Strip Drying Rack 0664442 Mfr # 49010510
COMPARE
Nobuto Filter Strip Drying Rack
₹24,500.00INR / Each