Advantec 800700 Nobuto Blood Filter Strip, 100/pk
Representative image only.
Advantec 800700 Nobuto Blood Filter Strip, 100/pk
Advantec | Mfr: 49010010| Item: 0664440
Ideal for collection applications
₹5,000.00INR / Pkg of 100
₹5,000.00INR / Pkg of 100
Buy It Together
Current Item
Advantec 800700 Nobuto Blood Filter Strip, 100/pk
Advantec 800700 Nobuto Blood Filter Strip, 100/pk
0664440
₹5,000.00INR / Pkg of 100
+
Advantec 800701 Nobuto Filter Strip Drying Rack
0664442
₹24,500.00INR / Each
=
Total Price 29500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Dried samples can be analyzed, stored, or mailed without refrigeration
  • 1 year warranty
More About this Item
Collect blood on the narrow end of the strip and place on the drying rack for easy and efficient sample handling. Each strip can hold approximately 0.1 mL of blood or 0.04 mL of serum. Strips are produced with high-purity cellulose paper with a fine, textile like structure—ideal for uniform sampling. Typical blood collection applications include toxoplasmosis (HA), hanta virus, new castles disease (HI), canine distemper (VN), measles (Rubeola), Japanese encephalitis (HI, VN), mycoplasmosis (AG), and hog cholera (VN).
Collect blood on the narrow end of the strip and place on the drying rack for easy and efficient sample handling. Each strip can hold approximately 0.1 mL of blood or 0.04 mL of serum. Strips are produced with high-purity cellulose paper with a fine, textile like structure—ideal for uniform sampling. Typical blood collection applications include toxoplasmosis (HA), hanta virus, new castles disease (HI), canine distemper (VN), measles (Rubeola), Japanese encephalitis (HI, VN), mycoplasmosis (AG), and hog cholera (VN).
Dried samples can be analyzed, stored, or mailed without refrigeration.
1 Year Warranty
Specifications
  • MaterialCellulose
  • DescriptionNobuto Blood Filter Strip, 100/pk