Advantec 800701 Nobuto Filter Strip Drying Rack
Representative image only.
Advantec 800701 Nobuto Filter Strip Drying Rack
Advantec | Mfr: 49010510| Item: 0664442
Ideal for collection applications
₹24,500.00INR / Each
₹24,500.00INR / Each
Product Description
Key Features
  • Dried samples can be analyzed, stored, or mailed without refrigeration
  • 1 year warranty
More About this Item
Collect blood on the narrow end of the strip and place on the drying rack for easy and efficient sample handling. Each strip can hold approximately 0.1 mL of blood or 0.04 mL of serum. Strips are produced with high-purity cellulose paper with a fine, textile like structure—ideal for uniform sampling. Typical blood collection applications include toxoplasmosis (HA), hanta virus, new castles disease (HI), canine distemper (VN), measles (Rubeola), Japanese encephalitis (HI, VN), mycoplasmosis (AG), and hog cholera (VN).
Simplify the drying and handling of Nobuto blood filter strips with this sturdy rack.
1 Year Warranty
Specifications
  • DescriptionNobuto Filter Strip Drying Rack