Advantec Hydrophobic PTFE Membranes (unsupported)
This thin, highly porous PTFE membrane is inert to most chemically aggressive solvents, strong acids, and bases that may be incompatible with supported PTFE filters. Ideal for sterilizing gases because it will trap aqueous aerosols on the surface. Operating temperature range: –120 to 260°C (–184 to 500°F).
46,490.00 - ₹96,670.00INR / Pkg of 100
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
Advantec T010A013A Unsupported Hydrophobic PTFE Membranes, 0.1 um, 13 mm; 100/Pk WW-36229-74 Mfr # T010A013A
COMPARE
13 Hydrophobic PTFE 0.1
Call For Price
Advantec T010A025A Unsupported Hydrophobic PTFE Membranes, 0.1 um, 25 mm; 100/Pk WW-36229-76 Mfr # T010A025A
COMPARE
25 Hydrophobic PTFE 0.1
Call For Price
Advantec T010A047A Unsupported Hydrophobic PTFE Membranes, 0.1 um, 47 mm; 100/Pk WW-36229-78 Mfr # T010A047A
COMPARE
47 Hydrophobic PTFE 0.1
Call For Price
Advantec T010A090C PTFE Hydrophobic Membranes (unsupported); 90 mm dia; 0.10 µm; 25/PK WW-36229-80 Mfr # T010A090C
COMPARE
90 Hydrophobic PTFE 0.1
Call For Price
Advantec T010A142C Unsupported Hydrophobic PTFE Membranes, 0.1 um, 142 mm; 25/Pk WW-36229-82 Mfr # T010A142C
COMPARE
142 Hydrophobic PTFE 0.1
Call For Price
Advantec T300A047A Unsupported Hydrophobic PTFE Membranes, 3.0 um, 47 mm; 100/Pk WW-36229-98 Mfr # T300A047A
COMPARE
47 Hydrophobic PTFE 3
Call For Price
Advantec T300A142C Unsupported Hydrophobic PTFE Membranes, 3.0 um, 142 mm; 25/Pk WW-36329-02 Mfr # T300A142C
COMPARE
142 Hydrophobic PTFE 3
₹82,750.00INR / Pkg of 25
Advantec T020A013A Unsupported Hydrophobic PTFE Membranes, 0.2 um, 13 mm; 100/Pk WW-36329-04 Mfr # T020A013A
COMPARE
13 Hydrophobic PTFE 0.2
₹46,490.00INR / Pkg of 100
Advantec T020A025A Unsupported Hydrophobic PTFE Membranes, 0.2 um, 25 mm; 100/Pk WW-36329-06 Mfr # T020A025A
COMPARE
25 Hydrophobic PTFE 0.2
₹58,300.00INR / Pkg of 100
Advantec T020A047A Unsupported Hydrophobic PTFE Membranes, 0.2 um, 47 mm; 100/Pk WW-36329-08 Mfr # T020A047A
COMPARE
47 Hydrophobic PTFE 0.2
₹79,860.00INR / Pkg of 100
Advantec T020A090C PTFE Hydrophobic Membranes (unsupported); 90 mm dia; 0.20 µm; 25/PK WW-36329-10 Mfr # T020A090C
COMPARE
90 Hydrophobic PTFE 0.2
₹63,670.00INR / Pkg of 25
Advantec T050A013A Unsupported Hydrophobic PTFE Membranes, 0.5 um, 13 mm; 100/Pk WW-36329-14 Mfr # T050A013A
COMPARE
13 Hydrophobic PTFE 0.5
₹46,490.00INR / Pkg of 100
Advantec T050A047A Unsupported Hydrophobic PTFE Membranes, 0.5 um, 47 mm; 100/Pk WW-36329-18 Mfr # T050A047A
COMPARE
47 Hydrophobic PTFE 0.5
₹74,020.00INR / Pkg of 100
Advantec T050A090C PTFE Hydrophobic Membranes (unsupported); 90 mm dia; 0.50 µm; 25/PK WW-36329-20 Mfr # T050A090C
COMPARE
90 Hydrophobic PTFE 0.5
₹63,670.00INR / Pkg of 25
Advantec T050A142C Unsupported Hydrophobic PTFE Membranes, 0.5 um, 142 mm; 25/Pk WW-36329-22 Mfr # T050A142C
COMPARE
142 Hydrophobic PTFE 0.5
₹82,750.00INR / Pkg of 25
Advantec T100A013A Unsupported Hydrophobic PTFE Membranes, 1.0 um, 13 mm; 100/Pk WW-36329-24 Mfr # T100A013A
COMPARE
13 Hydrophobic PTFE 1
₹46,490.00INR / Pkg of 100
Advantec T100A025A Unsupported Hydrophobic PTFE Membranes, 1.0 um, 25 mm; 100/Pk WW-36329-26 Mfr # T100A025A
COMPARE
25 Hydrophobic PTFE 1
₹58,300.00INR / Pkg of 100
Advantec T100A047A Unsupported Hydrophobic PTFE Membranes, 1.0 um, 47 mm; 100/Pk WW-36329-28 Mfr # T100A047A
COMPARE
47 Hydrophobic PTFE 1
₹74,020.00INR / Pkg of 100
Advantec T100A142C Unsupported Hydrophobic PTFE Membranes, 1.0 um, 142 mm; 25/Pk WW-36329-32 Mfr # T100A142C
COMPARE
142 Hydrophobic PTFE 1
₹82,750.00INR / Pkg of 25
Advantec T100A293D Unsupported Hydrophobic PTFE Membranes, 1.0 um, 293 mm; 25/Pk WW-36329-37 Mfr # T100A293D
COMPARE
293 Hydrophobic PTFE 1
₹96,670.00INR / Pkg of 25
Showing 1- 20 of 20