Adam Equipment Gladiator Industrial Washdown Scales
Find perfect balance of durability and performance for demanding applications
  • Backlit display changes color to swiftly notify of checkweighing limits
  • Keypad with color coding enhances efficiency
  • Preset tare function expedites repetitive weighing
  • Hold function allows weight reading to be held on screen
  • IP68-rated for washdown capabilities, protection against water and dust
Ruggedly designed to handle washdown weighing tasks in food, pharmaceutical and industrial applications. These scales feature easy-to-clean 304 stainless steel construction ideal for wet and dusty environments. Heavy-duty rubber feet optimize stability.
In addition to weighing and checkweighing, scales well-accommodate parts counting, percentage weighing, check counting and display hold applications. Programmable auto power-off helps to save energy, while external calibration allows for verification and adjustment with weights.
Available RS-232 interface enables quick connection with computers and printers.
282,000.00 - ₹427,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Readability (g) Description
Availability Pricing
Adam Equipment GGS16a Gladiator Industrial Washdown Scale, 16lb/8 kg  x 0.001lb/0.5g, 9.8" x 9.8" Platform 1113017 Mfr # GGS 16A
COMPARE
8 0.5 Gladiator Industrial Washdown Scale, 16lb/8 kg x 0.001lb/0.5g, 9.8" x 9.8" Platform
₹282,000.00INR / Each
Adam Equipment GGS 35A Gladiator Industrial Washdown Scale, 35lb/16kg  x 0.002lb/1g, 9.8" x 9.8" Platform 1113018 Mfr # GGS 35A
COMPARE
16 1 Gladiator Industrial Washdown Scale, 35lb/16kg x 0.002lb/1g, 9.8" x 9.8" Platform
Call For Price
Adam Equipment GGS 65A Gladiator Industrial Washdown Scale, 65lb/30kg  x 0.005lb/2g, 9.8" x 9.8" Platform 1113019 Mfr # GGS 65A
COMPARE
30 2 Gladiator Industrial Washdown Scale, 65lb/30kg x 0.005lb/2g, 9.8" x 9.8" Platform
₹330,000.00INR / Each
Adam Equipment GGB65a Gladiator Industrial Washdown Scale, 65lb/30kg  x 0.005lb/2g, 11.8" x 15.7" Platform 1113020 Mfr # GGB 65A
COMPARE
30 2 Gladiator Industrial Washdown Scale, 65lb/30kg x 0.005lb/2g, 11.8" x 15.7" Platform
₹308,000.00INR / Each
Adam Equipment GGB65aH Gladiator Industrial Washdown Scale, 65lb/30kg  x .0.002lb/1g, 11.8" x 15.7" Platform 1113021 Mfr # GGB 65AH
COMPARE
30 1 Gladiator Industrial Washdown Scale, 65lb/30kg x .0.002lb/1g, 11.8" x 15.7" Platform
₹399,000.00INR / Each
Adam Equipment GGB165a Gladiator Industrial Washdown Scale, 165lb/75kg  x 0.01lb/5g, 11.8" x 15.7" Platform 1113022 Mfr # GGB 165A
COMPARE
75 5 Gladiator Industrial Washdown Scale, 165lb/75kg x 0.01lb/5g, 11.8" x 15.7" Platform
₹308,000.00INR / Each
Adam Equipment GGF 165A Gladiator Industrial Washdown Scale, 165lb/75kg  x 0.01lb/5g, 15.7" x 19.7" Platform 1113023 Mfr # GGF 165A
COMPARE
75 5 Gladiator Industrial Washdown Scale, 165lb/75kg x 0.01lb/5g, 15.7" x 19.7" Platform
Call For Price
Adam Equipment GGF165aH Gladiator Industrial Washdown Scale, 165lb/75kg  x 0.005lb/2g, 15.7" x 19.7" Platform 1113024 Mfr # GGF 165AH
COMPARE
75 2 Gladiator Industrial Washdown Scale, 165lb/75kg x 0.005lb/2g, 15.7" x 19.7" Platform
₹427,000.00INR / Each
Adam Equipment GGF330a Gladiator Industrial Washdown Scale, 330lb/150kg  x 0.02lb/10g, 15.7" x 19.7" Platform 1113025 Mfr # GGF 330A
COMPARE
150 10 Gladiator Industrial Washdown Scale, 330lb/150kg x 0.02lb/10g, 15.7" x 19.7" Platform
₹391,000.00INR / Each
Adam Equipment GGL 330A Gladiator Industrial Washdown Scale, 330lb/150kg  x 0.02lb/10g, 17.7" x 23.6" Platform 1113026 Mfr # GGL 330A
COMPARE
150 10 Gladiator Industrial Washdown Scale, 330lb/150kg x 0.02lb/10g, 17.7" x 23.6" Platform
Call For Price
Showing 1- 10 of 10