Adam Equipment CPW Plus-P Industrial Scales (Piller Display)
These versatile scales are ideal for a wide variety of weighing tasks
  • Battery and AC operation
  • Splashproof keypad for use in wet environments
  • Hold function lets you freeze the reading on the display
  • Color-coded keypads with backlit display provide simple and quick operation
  • Steel base offers stability for weighing tasks
  • Removable stainless steel platform for easy cleaning
  • RS-232 interface
  • Pillar-mounted display
69,000.00 - ₹95,100.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Adam Equipment CPWPLUS 6P Industrial Scale with Column, 13 lb / 6 kg x 0.005 lb / 2 g, 115V 1112037 Mfr # CPWPLUS 6P
COMPARE
6 13 100 to 240
₹78,000.00INR / Each
Adam Equipment CPWPLUS 6P Industrial Scale with Column, 13Lb/6kg x 0.005Lb/2g, 230V 1112038 Mfr # CPWPLUS 6P-230V
COMPARE
6 13 230
Call For Price
Adam Equipment CPWPLUS 15P Industrial Scale, 33 lb / 15 kg x 0.01 lb / 5 g, 115V 1112039 Mfr # CPWPLUS 15P
COMPARE
15 33 100 to 240
₹80,100.00INR / Each
Adam Equipment CPWPLUS 15P Industrial Scale, 33Lb/15kg Capacity and 0.01Lb/5G Readability 230V 1112041 Mfr # CPWPLUS 15P-230V
COMPARE
15 33 230
Call For Price
Adam Equipment CPWPLUS 35P Industrial Scale, 75 lb / 35 kg x 0.02 lb / 10 g, 115V 1112042 Mfr # CPWPLUS 35P
COMPARE
35 75 100 to 240
₹81,500.00INR / Each
Adam Equipment CPWPLUS 35P Industrial Scale, 75Lb/35kg Capacity and 0.02Lb/10g Readability 230V 1112043 Mfr # CPWPLUS 35P-230V
COMPARE
35 75 230
Call For Price
Adam Equipment CPWPLUS 75P Industrial Scale with Column, 165 lb / 75 kg x 0.05 lb / 20 g, 115V 1112044 Mfr # CPWPLUS 75P
COMPARE
75 165 115
₹95,100.00INR / Each
Adam Equipment CPWPLUS 75P Industrial Scale with Column, 165Lb/75kg x 0.05Lb/20G, 230V 1112046 Mfr # CPWPLUS 75P-230V
COMPARE
75 165 230
Call For Price
Adam Equipment Industrial Scale with Column, 330 lb / 150 kg x 0.1 lb / 50 g, 115V 1112047 Mfr # CPWPLUS 150P
COMPARE
150 330 100 to 240
₹83,600.00INR / Each
Adam Equipment CPWPLUS 150P Industrial Scale with Column, 330lb/150kg x 0.1Lb/50G, 230V 1112048 Mfr # CPWPLUS 150P-230V
COMPARE
150 330 230
₹69,000.00INR / Each
Adam Equipment Industrial Scale with Column, 440 lb / 200 kg x 0.1 lb / 50 g, 115V 1112049 Mfr # CPWPLUS 200P
COMPARE
200 440 100 to 240
₹88,800.00INR / Each
Adam Equipment CPWPLUS 200P Industrial Scale with Column, 440lb/200kg x 0.1Lb/50G, 230V 1112050 Mfr # CPWPLUS 200P-230V
COMPARE
200 440 230
₹72,700.00INR / Each
Showing 1- 12 of 12