Adam Equipment CCT Cruiser Bench Counting Scales
Quick and accurate counting in a compact, rugged design
  • Three-window, backlit LCD provides quick results of weight, unit weight, and count
  • Solid ABS housing is durable for industrial use
  • Built-in memory stores products and settings
  • 1:600,000 internal counting resolution offers precise results
  • Preset tare function speeds process of repetitive weighing
  • Programmable auto power-off saves energy
  • Rechargeable battery lasts up to 90 hours on a single charge
These CCT Cruiser bench counting scales provide a quick and accurate method to check the number of pieces in a carton or container. User-friendly software keeps track of the count, automatically optimizing the unit weight. The weight can be displayed in pounds, grams, or kilograms. Color-coded keys allow for quick recognition of frequently used functions. Programmable backlit display can be customized to be always on, always off, or to light only when weighing.
Other features include automatic zero tracking, audible alarm for pre-set counts, automatic tare, pre-set tare, an accumulation facility that allows the count to be stored and recalled as an accumulated total. Supplied with a RS-232 bidirectional interface and real time clock.
147,000.00 - ₹176,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (kg) Readability (g)
Availability Pricing
Adam Equipment CCT 4 Cruiser Bench Counting Scale; 4 kg x 0.1 g 1180172 Mfr # CCT 4
COMPARE
External 4 0.1
₹147,000.00INR / Each
Adam Equipment CCT 8 Cruiser Bench Counting Scale; 8 kg x 0.2 g 1180173 Mfr # CCT 8
COMPARE
External 8 0.2
₹147,000.00INR / Each
Adam Equipment CCT 16 Cruiser Bench Counting Scale; 16 kg x 0.5 g 1180174 Mfr # CCT 16
COMPARE
External 16 0.5
₹147,000.00INR / Each
Adam Equipment CCT 32 Cruiser Bench Counting Scale; 32 kg  x 1 g 1180175 Mfr # CCT 32
COMPARE
External calibration 32 1
Call For Price
Adam Equipment CCT 48 Cruiser Bench Counting Scale; 48 kg x 2 g 1180176 Mfr # CCT 48
COMPARE
External 48 2
₹147,000.00INR / Each
Adam Equipment CCT 8UH Cruiser Bench Counting Scale; 8 kg x 0.05 g 1180177 Mfr # CCT 8UH
COMPARE
External 8 0.05
₹176,000.00INR / Each
Adam Equipment CCT 16UH Cruiser Bench Counting Scale; 16 kg x 0.1 g 1180178 Mfr # CCT 16UH
COMPARE
External 16 0.1
₹176,000.00INR / Each
Adam Equipment CCT 32UH Cruiser Bench Counting Scale; 32 kg x 0.2 g 1180179 Mfr # CCT 32UH
COMPARE
External calibration 32 0.2
Call For Price
Adam Equipment CCT 48UH Cruiser Bench Counting Scale; 48 kg x 0.5 g 1180180 Mfr # CCT 48UH
COMPARE
External 48 0.5
₹176,000.00INR / Each
Showing 1- 9 of 9