Accessories for Velp Vortex Mixers

These optional accessories accommodate a broad range of vortex mixers, greatly expanding their uses.
3,500.00 - ₹7,100.00INR / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Velp A00000012 Microtube foam Adapter for Vortex Mixers WW-86579-22 Mfr # A00000012
COMPARE
Microtube foam Adapter for Vortex Mixers
₹6,000.00INR / Each
Velp A00000015 Microplate foam Adapter, for Vortex Mixer WW-86579-23 Mfr # A00000015
COMPARE
Microplate foam Adapter, for Vortex Mixer
₹6,900.00INR / Each
Velp A00000014 16mm Test Tube foam Adapter, for Vortex Mixers. WW-86579-24 Mfr # A00000014
COMPARE
16mm Test Tube foam Adapter, for Vortex Mixers.
₹6,000.00INR / Each
Velp A00000016 Small rubber platform, 50mm, for Vortex Mixers. WW-86579-25 Mfr # A00000016
COMPARE
Small rubber platform, 50mm, for Vortex Mixers.
₹3,500.00INR / Each
Velp A00000017 Large foam platform, 94mm, for Vortex Mixers. WW-86579-26 Mfr # A00000017
COMPARE
Large foam platform, 94mm, for Vortex Mixers.
₹4,000.00INR / Each
Velp A00000013 Large soft foam platform, 94mm, for Vortex Mixers. WW-86579-27 Mfr # A00000013
COMPARE
Large soft foam platform, 94mm, for Vortex Mixers.
₹4,800.00INR / Each
Velp 10002021 Replacement Pop-off cup, for use with Wizard Vortex Mixer WW-86579-28 Mfr # 10002021
COMPARE
Replacement Pop-off cup, for use with Wizard Vortex Mixer
₹7,100.00INR / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat