Velp A00000013 Large soft foam platform, 94mm, for Vortex Mixers.

Representative image only.

Velp A00000013 Large soft foam platform, 94mm, for Vortex Mixers.

Velp | Mfr: A00000013| Item: 8657927

₹5,800.00INR / Each

₹5,800.00INR / Each

Buy It Together

Current Item

Velp A00000013 Large soft foam platform, 94mm, for Vortex Mixers.
Velp A00000013 Large soft foam platform, 94mm, for Vortex Mixers.
8657927

₹5,800.00INR / Each
+ + =
Total Price 133810.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

86579, 86579-2

More About this Item

These optional accessories accommodate a broad range of vortex mixers, greatly expanding their uses.

Soft Foam, 3 3/4" (9.5 cm) dia.

Conformite Europeenne
Specifications
  • DescriptionLarge soft foam platform, 94mm, for Vortex Mixers.
Reviews
Q&A