Velp F202A0270 TX4 Digital Vortex Mixer with Infra-Red Sensor, 100-240 V
Representative image only.
Velp F202A0270 TX4 Digital Vortex Mixer with Infra-Red Sensor, 100-240 V
Velp | Mfr: F202A0270| Item: 8657904
Digital control of speed and time for high repeatability
₹68,270.00INR / Each
₹68,270.00INR / Each
Buy It Together
Current Item
Velp F202A0270 TX4 Digital Vortex Mixer with Infra-Red Sensor, 100-240 V
Velp F202A0270 TX4 Digital Vortex Mixer with Infra-Red Sensor, 100-240 V
8657904
₹68,270.00INR / Each
+
Velp A00000012 Microtube foam Adapter for Vortex Mixers
8657922
₹7,400.00INR / Each
=
Total Price 75670.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
standard pop-off cup for single test tube and a 6-ft (1.8-m) power cord with plug.
Key Features
  • Unique infrared sensor drastically reduces repetitive strain on your wrists, or operate in continuous “touch” mode
  • Variable speeds up to 3000 rpm for applications from gentle mixing to vigorous vortexing
  • Large, clear display shows elapsed time and speed
  • Timer can be set up to 999:59 minutes; unit will shut off when session is completed
  • Quiet operation with soft-start feature and controlled ramping
  • Polymer/zinc alloy construction is easy to clean and offers excellent chemical resistance
  • Four anti-sliding feet provide enhanced stability on many surface types
  • Optional accessories accommodate a broad range of vessels, greatly expanding the uses
  • 2 year warranty
Two year warrantyConformite Europeenne
Specifications
  • Power (VAC)100 to 240
  • DescriptionTX4 Digital Vortex Mixer with Infra-Red Sensor, 100-240 VAC.