Velp A00000017 Large foam platform, 94mm, for Vortex Mixers.

Representative image only.

Velp A00000017 Large foam platform, 94mm, for Vortex Mixers.

Velp | Mfr: A00000017| Item: 8657926

₹4,700.00INR / Each

₹4,700.00INR / Each

Buy It Together

Current Item

Velp A00000017 Large foam platform, 94mm, for Vortex Mixers.
Velp A00000017 Large foam platform, 94mm, for Vortex Mixers.
8657926

₹4,700.00INR / Each
+ + =
Total Price 132710.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

86579, 86579-2

More About this Item

These optional accessories accommodate a broad range of vortex mixers, greatly expanding their uses.

Foam, 3 3/4" (9.5 cm) dia.

Specifications
  • DescriptionLarge foam platform, 94mm, for Vortex Mixers.
Reviews
Q&A