Accessories for Boekel Scientific Tube Rotator

Use with Boekel Scientific Tube Rotator.
47,000.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Boekel Scientific 260651 Rotor Attachment holds 1.5 - 2.0 ml tubes 5120205 Mfr # 260651
COMPARE
Rotor Attachment holds 1.5 - 2.0 ml tubes
₹47,000.00INR / Each
Boekel Scientific 260652 Rotor Attachment holds 15 ml tubes 5120206 Mfr # 260652
COMPARE
Rotor Attachment holds 1.5 ml tubes
₹47,000.00INR / Each
Boekel Scientific 260653 Rotor Attachment holds 50 ml tubes 5120207 Mfr # 260653
COMPARE
Rotor Attachment holds 50 ml tubes
₹47,000.00INR / Each
Boekel Scientific 260654 Rotor Attachment holds 18.5mm culture tubes 5120208 Mfr # 260654
COMPARE
Rotor Attachment holds 18-5mm culture tubes
₹47,000.00INR / Each
Boekel Scientific 260655 Rotor Attachment holds 14mm culture tubes 5120209 Mfr # 260655
COMPARE
Rotor Attachment holds 14mm culture tubes
₹47,000.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat