Boekel Scientific 260652 Rotor Attachment holds 15 ml tubes

Representative image only.

Boekel Scientific 260652 Rotor Attachment holds 15 ml tubes

Boekel Scientific | Mfr: 260652| Item: 5120206

₹47,000.00INR / Each

₹47,000.00INR / Each

Buy It Together

Current Item

Boekel Scientific 260652 Rotor Attachment holds 15 ml tubes
Boekel Scientific 260652 Rotor Attachment holds 15 ml tubes
5120206

₹47,000.00INR / Each
+ +
Boekel Scientific 260650 Tube Rotator, 100-240V
5120200

₹364,000.00INR / Each
+ =
Total Price 619650.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Rotor attachment for Boekel Scientific Tube Rotator 51202-00
  • 2 year warranty

More About this Item

Use with Boekel Scientific Tube Rotator.

Two year warranty
Specifications
  • DescriptionRotor Attachment holds 1.5 ml tubes
Reviews
Q&A