A&D Weighing SJ Portion Scales
Fast stability time – Less than 1 second
  • Stainless steel weighing pan
  • High resolution (SJ-5000HS:1/2500, SJ-12KHS:1/2400, SJ-1000HS/2000HS :1/2000)
  • Large wide LCD display (25mm height)
  • Durable body
  • Large weighing pan 230mm x 190mm
  • Full digital calibration function
  • Multiple weighing units – g, lb, oz and lb-oz
  • Battery operation with 4 x D size dry batteries
  • Simple and easy to change the batteries
  • Optional AC adapter
  • Automatic power OFF function
  • Spirit level and adjustable feet for accurate weighing
  • Status indication function – Battery, Stable, Zero and Net
52,200.00 - ₹62,500.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Capacity (kg) Readability (g)
Availability Pricing
A&D Weighing SJ-1000HSF Digital Portion Scale, 2.2Lb x 0.001Lb 1111121 Mfr # SJ-1000HSF
COMPARE
1000 1 0.5
₹52,200.00INR / Each
A&D Weighing SJ-2000HSF Digital Portion Scale, 4.4Lb x 0.002Lb 1111122 Mfr # SJ-2000HSF
COMPARE
2000 2 1
₹52,200.00INR / Each
A&D Weighing SJ-5000HSF Digital Portion Scale, 11Lb x 0.005Lb 1111123 Mfr # SJ-5000HSF
COMPARE
5000 5 2
₹52,200.00INR / Each
A&D Weighing SJ-5001HSF Digital Portion Scale,  11Lb x 0.005Lb 1111124 Mfr # SJ-5001HSF
COMPARE
5000 5 2
₹57,300.00INR / Each
A&D Weighing SJ-12KHSF Digital Portion Scale, 26Lb x 0.01Lb 1111125 Mfr # SJ-12KHSF
COMPARE
12000 12 5
₹52,200.00INR / Each
A&D Weighing SJ-20KHSF Digital Portion Scale, 44Lb x 0.02Lb 1111126 Mfr # SJ-20KHSF
COMPARE
20000 20 10
₹61,500.00INR / Each
A&D Weighing SJ-30KHSF Digital Portion Scale, 66Lb x 0.05Lb 1111127 Mfr # SJ-30KHSF
COMPARE
30000 30 20
₹62,500.00INR / Each
Showing 1- 7 of 7