A&D Everest Eki Portable Balances
Compact, lightweight body for easy portability
  • Keypad calibration with external weight
  • Back light for use in low-light conditions
  • Rechargeable battery provides over eight hours of use on one charge
  • Parts counting, percent weighing, HI/OK/LO comparator, remote zeroing, and full-scale tare
116,000.00 - ₹213,000.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
A&D Weighing EK-410i Compact Balance 400g x 0.01g Counting, Percentage, Comaparator Modes 1111025 Mfr # EK-410I
COMPARE
400 115 0.01
₹159,000.00INR / Each
A&D Weighing EK-610i Portable Balance, 600g x 0.01g 1111026 Mfr # EK-610I
COMPARE
600 115 0.01
₹187,000.00INR / Each
A&D Weighing EK-4100i Portable Balance, 4000g x 0.1g 1111027 Mfr # EK-4100I
COMPARE
4000 115 0.1
₹175,000.00INR / Each
A&D Weighing EK-6100i Portable Balance, 6000g x 0.1g 1111028 Mfr # EK-6100I
COMPARE
6000 115 0.1
₹213,000.00INR / Each
A&D Weighing EK-120I Portable Balance, 120g x 0.01g 1111900 Mfr # EK-120I
COMPARE
120 115 0.01
₹126,000.00INR / Each
A&D Weighing EW-150I Portable Balance, 150g x 0.05g 1111905 Mfr # EW-150I
COMPARE
150 120 0.05
Call For Price
A&D Weighing EK-200I Portable Balance, 200g x 0.01g 1111910 Mfr # EK-200I
COMPARE
200 115 0.01
₹135,000.00INR / Each
A&D Weighing EK-300I Portable Balance, 300g Capacity 1111915 Mfr # EK-300I
COMPARE
300 115 0.01
₹142,000.00INR / Each
A&D Weighing EK-600I Portable Balance, 600g Capacity 1111920 Mfr # EK-600I
COMPARE
600 115 0.1
₹124,000.00INR / Each
A&D Weighing EK-1200I Portable Balance, 1200g Capacity 1111925 Mfr # EK-1200I
COMPARE
1200 115 0.1
₹116,000.00INR / Each
A&D Weighing EK-2000I Portable Balance, 2000g Capacity 1111935 Mfr # EK-2000I
COMPARE
2000 115 0.1
₹135,000.00INR / Each
A&D Weighing EK-3000I Portable Balance, 3000g Capacity 1111940 Mfr # EK-3000I
COMPARE
3000 115 0.1
₹147,000.00INR / Each
A&D Weighing EK-6000I Portable Balance, 6000g Capacity 1111945 Mfr # EK-6000I
COMPARE
6000 115 1
₹139,000.00INR / Each
A&D Weighing EK-12KI Portable Balance, 12000g Capacity 1111950 Mfr # EK-12KI
COMPARE
12000 115 1
₹171,000.00INR / Each
A&D Weighing EW-12KI Portable Balance, 12000g Capacity 1111955 Mfr # EW-12KI
COMPARE
12000 120 5
Call For Price
Showing 1- 15 of 15