Watson-Marlow 318MCX Multichannel Microcassette Extension Pump Head, 5-Channel, 8-Roller
Representative image only.
Watson-Marlow 318MCX Multichannel Microcassette Extension Pump Head, 5-Channel, 8-Roller
Watson-Marlow | Mfr: 033.6854.000| Item: 5013019
Specially designed for low-flow applications
₹110,000.00INR / Each
₹110,000.00INR / Each
Buy It Together
Current Item
Product Description
Key Features
  • Built-in tubing stops ensure correct position and tension
  • Two-segmented tubing sections extend tubing life
  • Remove cassette without disturbing other channels
  • Get up to 10 channels with an extension pump head
  • 5-channel, 8-roller extension pump head
  • Extension pump head adds five more channels
  • Eight-roller model is ideal for applications requiring higher precision and less pulsation
  • 1 year warranty
More About this Item
Use the multichannel microcassette pump heads to achieve a flow rate as low as 0.002 mL/min per channel. Each pump head comes with five pumping channels—select the four-roller model for higher flow rate applications or the eight-roller model for higher precision with less pulsation. Each cassette loads easily with a single cam lever action. Add an extension pump head to double your pumping capability to 10 channels.
These peristaltic pump heads accept tubing with built-in stops that ensure the correct position and tension of the tubing. Easily identify tubing size by the color of the stops. Each tubing piece comes with three stops that divide the tubing into two pumping sections, doubling the pump life of the tubing. Preload the tubing into the cassette before placing it into the pump head to not disturb other channels being used.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
1 Year Warranty
Specifications
  • Series300
  • Number of Channels5
  • Max Flow Rate Per Channel (mL/min)36
  • Min Flow Rate Per Channel (mL/min)0.002
  • Pump Head318MCX
  • Tubing sizes accepted2-stop tubing with the following bore sizes: 0.13, 0.19, 0.25, 0.38, 0.5, 0.63, 0.76, 0.88, 1.02, 1.14, 1.29, 1.42, 1.52, 1.65, 1.85, 2.05, 2.29, 2.54, 2.79 mm
  • Included AccessoriesPump head with 5 cassettes
  • Number of Rollers8
  • Housing MaterialAluminum
  • Rotor MaterialAluminum
  • DescriptionMultichannel Microcassette Extension Pump Head, 5-Channel, 8-Roller