Watson-Marlow 300-Series Multichannel Microcassette Pump Heads
Specially designed for low-flow applications
  • Built-in tubing stops ensure correct position and tension
  • Two-segmented tubing sections extend tubing life
  • Remove cassette without disturbing other channels
  • Get up to 10 channels with an extension pump head
Use the multichannel microcassette pump heads to achieve a flow rate as low as 0.002 mL/min per channel. Each pump head comes with five pumping channels—select the four-roller model for higher flow rate applications or the eight-roller model for higher precision with less pulsation. Each cassette loads easily with a single cam lever action. Add an extension pump head to double your pumping capability to 10 channels.
These peristaltic pump heads accept tubing with built-in stops that ensure the correct position and tension of the tubing. Easily identify tubing size by the color of the stops. Each tubing piece comes with three stops that divide the tubing into two pumping sections, doubling the pump life of the tubing. Preload the tubing into the cassette before placing it into the pump head to not disturb other channels being used.
103,000.00 - ₹110,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Channel (mL/min) Number of Rollers Description
Availability Pricing
Watson-Marlow 314MC Multichannel Microcassette Pump Head, 5-Channel, 4-Roller 5013016 Mfr # 033.6453.000
COMPARE
53 4 Multichannel Microcassette Pump Head, 5-Channel, 4-Roller
₹103,000.00INR / Each
Watson-Marlow 314MCX Multichannel Microcassette Extension Pump Head, 5-Channel, 4-Roller 5013017 Mfr # 033.6454.000
COMPARE
53 4 Multichannel Microcassette Extension Pump Head, 5-Channel, 4-Roller
₹103,000.00INR / Each
Watson-Marlow 318MC Multichannel Microcassette Pump Head, 5-Channel, 8-Roller 5013018 Mfr # 033.6853.000
COMPARE
36 8 Multichannel Microcassette Pump Head, 5-Channel, 8-Roller
₹110,000.00INR / Each
Watson-Marlow 318MCX Multichannel Microcassette Extension Pump Head, 5-Channel, 8-Roller 5013019 Mfr # 033.6854.000
COMPARE
36 8 Multichannel Microcassette Extension Pump Head, 5-Channel, 8-Roller
₹110,000.00INR / Each