Watson-Marlow 120U/DM3 Variable-Speed Pump System, Manual/Analog Control, 0.1 to 200 rpm, Three-Channel, 4-Roller; 90 to 264 VAC
Representative image only.
Watson-Marlow 120U/DM3 Variable-Speed Pump System, Manual/Analog Control, 0.1 to 200 rpm, Three-Channel, 4-Roller; 90 to 264 VAC
Watson-Marlow | Mfr: 010.6141.M30| Item: 5014200
Compact pump systems with remote control features deliver accuracy and repeatability to optimize your fluid handling process
₹1,114,000.00INR / Each
for an additional
₹37,400.00INR / Each
for an additional
₹38,500.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹1,114,000.00INR / Each
Buy It Together
Current Item
Watson-Marlow 120U/DM3 Variable-Speed Pump System, Manual/Analog Control, 0.1 to 200 rpm, Three-Channel, 4-Roller; 90 to 264 VAC
Watson-Marlow 120U/DM3 Variable-Speed Pump System, Manual/Analog Control, 0.1 to 200 rpm, Three-Channel, 4-Roller; 90 to 264 VAC
5014200
₹1,114,000.00INR / Each
+ + +
InnoCal NIST-Traceable Calibration; Peristaltic Pump Drive
1705001
₹37,400.00INR / Each
+ + + + + + =
Total Price 1477290.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Remotely control start/stop, speed, and pump direction
  • Small, stackable pumps require minimal benchtop space
  • Control and view speed on digital LCD
  • Keypad lock prevents setting accidental changes
  • Smooth IP31-rated housing for easy wipedown
  • Auto key for changing between manual and auto control
  • 3 year warranty
More About this Item
Easily maintain fluid integrity using peristaltic pumps. Only the bore of the tubing contacts the fluid, eliminating the risk of the pump contaminating the fluid or the fluid contaminating the pump. Peristaltic pumps are great for use with most liquids, including viscous, shear-sensitive, corrosive, and abrasive fluids.
Manually or remotely control these compact pumps for accurate and repeatable fluid transfer. Each pump system includes a single-channel pump head to deliver flow rates from 0.001 to 170 mL/min*—select from 2-roller, 4-roller, or 4-roller flip-top models. Simple manual control allows start/stop, direction change, and speed adjustment from the keypad; adjust speed in 0.1 rpm increments. Press the MAX key for rapid priming; AUTO START for automatic power recovery. Remotely control start/stop, pump direction, and speed using analog signals (4 to 20 mA, 0 to 10 V); 0 to 5 V output allows remote speed display.
Use the keypad lock feature to prevent any unwanted changes to pump settings. A smooth, IP31-rated housing encases a maintenance-free, brushless motor for quick and easy cleaning. Contact us for free technical application support on pump & tubing selection.
*Flow rate depends on drive rpm, pump head, and tubing size.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Conformite EuropeenneCanadian Standards Association3 Year Warranty
Specifications
  • Series100
  • Control TypeManual, analog
  • Min RPM0.1
  • Max RPM200
  • Number of Channels3
  • Max Flow Rate (mL/min)36
  • Min Flow Rate (mL/min)0.0001
  • Drive120U
  • Pump Head400DM3 four-roller
  • Included AccessoriesDrive with universal plug attachments and pump head
  • Drive IP ratingIP 31
  • Number of Rollers4
  • Max Pressure (PSI)30
  • Power (VAC)90 to 264
  • Power (Hz)47 to 63 Hz
  • Housing MaterialABS plastic
  • DescriptionVariable-Speed Pump System, Manual/Analog Control, 0.1 to 200 rpm, Three-Channel, 4-Roller; 90 to 264 VAC
  • Warranty3 years