Cole-Parmer® Food, Milk, and Dairy Tubing
Smooth, nonporous bore inhibits particle entrapment
  • Crystal clear for visual inspection and flow monitoring
  • Non-toxic, odor-free, and taste-free
  • Smooth, non-porous and flexible tubing promotes a sanitary fluid path
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Cole-Parmer® Food, Milk, and Dairy Tubing is non-toxic, taste-free and odor-free, and meets all applicable regulatory standards for use in food, dairy and milk applications. Producers of food, milk and dairy products require the dependable performance of this tubing in countless filling, draining, transfer and processing applications. Smooth, non-porous and flexible, the Food, Milk, and Dairy tubing promotes a sanitary fluid path by maintaining an interior free of trapped particles, and minimizing potential for bacterial growth. In addition, it is resistant to harsh alkaline cleaners and is not affected by most commonly used sanitizers. Tubing has been sanctioned by the US Food & Drug Administration for food applications under Title 21 of the Code of Federal Regulations. Tubing material is 3A compliant.
2,300.00 - ₹301,000.00INR / Pkg of 1
34 variations of this product are available.
See All Product Options Below
34 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/16 1/8 1/32
₹2,300.00INR / Pkg of 1
COMPARE
3/32 5/32 1/32
₹3,000.00INR / Pkg of 1
COMPARE
1/8 3/16 1/32
₹3,600.00INR / Pkg of 1
COMPARE
1/8 1/4 1/16
₹8,700.00INR / Pkg of 1
COMPARE
3/16 5/16 1/16
₹12,000.00INR / Pkg of 1
COMPARE
3/16 1/4 1/32
₹5,000.00INR / Pkg of 1
COMPARE
3/16 7/16 1/8
₹26,500.00INR / Pkg of 1
COMPARE
1/4 3/8 1/16
₹14,600.00INR / Pkg of 1
COMPARE
1/4 7/16 3/32
₹23,500.00INR / Pkg of 1
COMPARE
1/4 1/2 1/8
₹28,500.00INR / Pkg of 1
COMPARE
5/16 7/16 1/16
₹16,900.00INR / Pkg of 1
COMPARE
5/16 1/2 3/32
₹24,600.00INR / Pkg of 1
COMPARE
5/16 9/16 1/8
₹29,600.00INR / Pkg of 1
COMPARE
3/8 1/2 1/16
₹19,600.00INR / Pkg of 1
COMPARE
3/8 9/16 3/32
₹25,300.00INR / Pkg of 1
COMPARE
3/8 5/8 1/8
₹36,600.00INR / Pkg of 1
COMPARE
7/16 9/16 1/16
₹23,200.00INR / Pkg of 1
COMPARE
1/2 5/8 1/16
₹23,800.00INR / Pkg of 1
COMPARE
1/2 11/16 3/32
₹33,000.00INR / Pkg of 1
COMPARE
1/2 3/4 1/8
₹45,200.00INR / Pkg of 1
Showing 1 - 20 of 34