Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Slip x 200 Series Barb, 1/8" ID; 25/Pk
Representative image only.
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Slip x 200 Series Barb, 1/8" ID; 25/Pk
Value Plastics | Mfr: MTLS230-1| Item: 5012079
A secure connection under pressure with 200 series barbs
₹2,400.00INR / Pkg of 25
₹2,400.00INR / Pkg of 25
Buy It Together
Current Item
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Slip x 200 Series Barb, 1/8" ID; 25/Pk
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Slip x 200 Series Barb, 1/8" ID; 25/Pk
5012079
₹2,400.00INR / Pkg of 25
+
Cole-Parmer TPE Tubing, Black, 1/8" ID x 3/8" OD; 25 Ft
5012133
₹6,400.00INR / Pkg of 1
=
Total Price 8800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Barbed ridges provide secure grip on the inside of flexible tubing
  • Slip luer fittings allow for a simple yet secure connection
  • Constructed of durable chemical resistance nylon
More About this Item
The Value Plastics Male Luer Slip to 200 Series Barb are designed to provide outstanding performance characteristics when used with flexible tubing. Tubing is expanded by 50% above its nominal inside diameter, permitting high-pressure capabilities. The male luer slip connection ensures a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. Adapters are available in various materials to accommodate various applications and compliance requirements.
White nylon is suitable for Gamma radiation, ethylene oxide (EtO), and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Width (InchShort)3/8
  • Height (InchShort)57/64
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Fitting Width (in)3/8
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;RoHS
  • Luer Connection Type (Port 1)Male Luer Slip
  • Luer Connection Type (Port 2)Hose Barb
  • DescriptionLuer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Slip x 200 Series Barb, 1/8" ID; 25/Pk