Value Plastics Male Luer Slip to 200 Series Barb
A secure connection under pressure with 200 series barbs
  • Quick and easy assembly and disassembly without the need for tools
  • Barbed ridges provide secure grip on the inside of flexible tubing
  • Slip luer fittings allow for a simple yet secure connection
The Value Plastics Male Luer Slip to 200 Series Barb are designed to provide outstanding performance characteristics when used with flexible tubing. Tubing is expanded by 50% above its nominal inside diameter, permitting high-pressure capabilities. The male luer slip connection ensures a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. Adapters are available in various materials to accommodate various applications and compliance requirements.
2,400.00 - ₹3,200.00INR / Pkg of 25
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Regulatory Approvals
Availability Pricing
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Slip x 200 Series Barb, 1/16" ID; 25/Pk 5012076 Mfr # MTLS210-1
COMPARE
1/16 Nylon Non-Phthalate
₹2,400.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Slip x 200 Series Barb, 1/16" ID; 25/Pk 5012077 Mfr # MTLS210-6005
COMPARE
1/16 Polypropylene Non-Phthalate
₹3,200.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Slip x 200 Series Barb, 3/32" ID; 25/Pk 5012078 Mfr # MTLS220-6005
COMPARE
3/32 Polypropylene ADCF
₹3,200.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Slip x 200 Series Barb, 1/8" ID; 25/Pk 5012079 Mfr # MTLS230-1
COMPARE
1/8 Nylon Non-DEHP
₹2,400.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Slip x 200 Series Barb, 1/8" ID; 25/Pk 5012080 Mfr # MTLS230-6005
COMPARE
1/8 Polypropylene ADCF
₹3,200.00INR / Pkg of 25