Traceable Robo™ Thermometer Ultra; ±0.4°C Accuracy at Tested Points

Representative image only.

Traceable Robo™ Thermometer Ultra; ±0.4°C Accuracy at Tested Points

Traceable | Mfr: 4349TR| Item: 9876735

180° rotating display lets you view readings from any angle

₹11,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹11,000.00INR / Each

Buy It Together

Current Item

Traceable Robo™ Thermometer Ultra; ±0.4°C Accuracy at Tested Points
Traceable Robo™ Thermometer Ultra; ±0.4°C Accuracy at Tested Points
9876735

₹11,000.00INR / Each
+ + + + + + + =
Total Price 143500.00 ADD 8 ITEMS TO CART
Product Description

Includes

Traceable® Certificate, probe case with clip, and battery.

Key Features

  • Measure liquids and semisolids with the piercing-tip probe
  • Extra long 8” (200 mm) stem is ideal for measuring reagents, water baths, air and liquids
  • °C/°F switchable
  • ±0.4°C accuracy at tested points
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty

More About this Item

Easily view temperature readings from any angle with the 180° rotating display. Thermometer features an 8" long, piercing-tip probe—ideal for measuring both liquids and semisolids in water baths, vats, and deep containers.

Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.

Guaranteed to have at least one year of calibration remaining on certificate.

1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Specifications
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° F)-58
  • Max Temperature (° F)536
  • Min Temperature (° C)-50
  • Max Temperature (° C)280
  • Accuracy±0.4°C (±0.7°F) at tested points; otherwise ±1.0°C (±1.8°F)
  • Resolution0.1° (above –19.9° and below 199.9°); otherwise 1°C
  • Display Type3.5-digit LCD, 1/4"H digits
  • Width (in)2
  • Length (in)9 1/4
  • No. of Probe(s) Included1
  • Probe Diameter (in)0.14
  • Probe Length (in)8
  • Battery1 1.5V Silver Oxide (included)
  • DescriptionThermometer Ultra; ±0.4°C Accuracy at Tested Points
  • Warranty1 year
Reviews
Q&A