Traceable® Robo™ Thermometers with Calibration
180° rotating display lets you view readings from any angle
  • Measure liquids and semisolids with the piercing-tip probe
  • Extra long 8” (200 mm) stem is ideal for measuring reagents, water baths, air and liquids
  • °C/°F switchable
Easily view temperature readings from any angle with the 180° rotating display. Thermometer features an 8" long, piercing-tip probe—ideal for measuring both liquids and semisolids in water baths, vats, and deep containers.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
11,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy
Availability Pricing
Traceable Robo™ Thermometer Ultra; ±0.4°C Accuracy at Tested Points 9876735 Mfr # 4349TR
COMPARE
±0.4°C (±0.7°F) at tested points; otherwise ±1.0°C (±1.8°F)
₹11,000.00INR / Each