Traceable 6603 Replacement Smart Stainless Steel Probe for 18004-37
Representative image only.
Traceable 6603 Replacement Smart Stainless Steel Probe for 18004-37
Traceable | Mfr: 6603CC| Item: 1800438
Save time and money by simply replacing probes that are damaged or due for calibration
₹53,100.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹53,100.00INR / Each
Buy It Together
Current Item
Traceable 6603 Replacement Smart Stainless Steel Probe for 18004-37
Traceable 6603 Replacement Smart Stainless Steel Probe for 18004-37
1800438
₹53,100.00INR / Each
+ + =
Total Price 132800.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Thermometer recalibration accomplished by simply replacing the probe
  • Durable 10-ft (3-m) long microcable
  • Durable 10-ft (3-m) long microcable
  • Stainless steel probe
  • Use with thermometer 18004-37
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
More About this Item
Calibration data are stored in the probe, so simply replace the probe when it is time for recalibration. Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
These replacement temperature probes are compatible with the Traceable® Data Logging Ethernet Thermometers. Each probe comes with a 10-ft (3-m) long microcable that withstands refrigerator/freezer door closing on it. Rugged stainless steel probe withstands a wide temperature range and is suitable for use with the Traceable 6601 Wide-Range Data Logging Ethernet Thermometer
1 Year WarrantyNIST Traceable Calibration
Specifications
  • Probe TypePtRTD
  • Min Temperature (° F)-130
  • Max Temperature (° F)221
  • Min Temperature (° C)-90
  • Max Temperature (° C)105
  • Sheath MaterialStainless Steel
  • Compatible WithEthernet Smart Data Logging Wide-Range Thermometer 18004-37
  • DescriptionReplacement Smart Stainless Steel Probe for 18004-37