Thermo Scientific SureSTART™ Certified Snap Cap, 11 mm, Level 2, Polyethylene, White Virgin PTFE Septum; 100/Pk
Representative image only.
Thermo Scientific SureSTART™ Certified Snap Cap, 11 mm, Level 2, Polyethylene, White Virgin PTFE Septum; 100/Pk
Thermo Scientific | Mfr: 6ARC11-P| Item: 9881212
Keep your samples safe with these vial closures
₹6,500.00INR / Pkg of 100
₹6,500.00INR / Pkg of 100
Buy It Together
Current Item
Thermo Scientific SureSTART™ Certified Snap Cap, 11 mm, Level 2, Polyethylene, White Virgin PTFE Septum; 100/Pk
Thermo Scientific SureSTART™ Certified Snap Cap, 11 mm, Level 2, Polyethylene, White Virgin PTFE Septum; 100/Pk
9881212
₹6,500.00INR / Pkg of 100
+ + + + + =
Total Price 92300.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Certified performance caps are quality tested to deliver the reproducibility you need
  • Decrease your risk of failed analysis and sample reruns
  • Easily secure vial contents with screw, snap, or crimp caps
More About this Item
Meet your certification and documentation needs with the Thermo Scientific™ SureSTART™ specification certified vial caps. Level 1 products are cost optimized, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. Level 2 products have very low metal contents, extractables, and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. Level 3 products are packed in clean rooms to protect against environmental factors. They contain the lowest levels of metals, extractables, and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes.
Caps are available in different closure types, colors, and septum materials to suit your application. AVCS™ screw caps incorporate Advanced Vial Closure System technology to ensure optimal seal compression when closing a vial. Snap caps can be snapped on and off by hand which eliminates the need for metal crimping and decapping tools. Crimp caps crimps tightly around the neck for a tight seal.
Specifications
  • Cap TypeSnap Cap
  • Cap Diameter (mm)11
  • Cap MaterialPolyethylene
  • Cap colorClear
  • Septum TypeWhite Virgin PTFE
  • Level2
  • DescriptionCertified Snap Cap, 11 mm, Level 2, Polyethylene, White Virgin PTFE Septum; 100/Pk