Thermo Scientific™ SureSTART™ EPA Certified Screw Vial and Cap Kits, Level 2
Vial kits combine vials and caps with septa for your convenience
  • Certified level 2 EPA vials are ideal for high-throughput analysis
  • Septa are manufactured for contaminant free sampling
  • Processed and packaged under strict ISO standards
Ensure that your environmental analyses meet government regulations with the Level 2 Thermo Scientific™ SureSTART™ EPA certified screw vials and caps. These convenience kits contain glass screw vials and white, open-top, polypropylene caps with a range of septa options.
Level 2 products have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow. EPA vials are processed and packaged under a strict registered ISO Quality Management System. Every case of product is labeled with its production number and is custody sealed to ensure reliable chain-of-custody.
15,300.00 - ₹45,300.00INR / Pkg of 100
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Septum Type Tube Color Volume (mL)
Availability Pricing
Thermo Scientific SureSTART™ Vial Kit, Clear Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 4 mL; 100/Pk 0892222 Mfr # 6AK4CCT
COMPARE
Polyethylene foam/PTFE Clear 4
₹15,300.00INR / Pkg of 100
Thermo Scientific SureSTART™ Vial Kit, Clear Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 8 mL; 200/Pk 0892223 Mfr # 6AK8CCT
COMPARE
Polyethylene foam/PTFE Clear 8
₹30,000.00INR / Pkg of 200
Thermo Scientific SureSTART™ Vial Kit, Clear Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 12 mL; 200/Pk 0892224 Mfr # 6AK12CCT
COMPARE
Polyethylene foam/PTFE Clear 12
₹40,700.00INR / Pkg of 200
Thermo Scientific SureSTART™ Vial Kit, Clear Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 16 mL; 200/Pk 0892226 Mfr # 6AK16CCT
COMPARE
Polyethylene foam/PTFE Clear 16
₹43,800.00INR / Pkg of 200
Thermo Scientific SureSTART™ Vial Kit, Clear Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 20 mL; 100/Pk 0892227 Mfr # 6AK20CCTAS
COMPARE
Polyethylene foam/PTFE Clear 20
₹23,100.00INR / Pkg of 100
Thermo Scientific SureSTART™ Vial Kit, Clear Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 40 mL; 100/Pk 0892229 Mfr # 6AK40CCT
COMPARE
Polyethylene foam/PTFE Clear 40
₹27,600.00INR / Pkg of 100
Thermo Scientific SureSTART™ Vial Kit, Amber Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 4 mL; 100/Pk 0892232 Mfr # 6AK4ACT
COMPARE
Polyethylene foam/PTFE Amber 4
₹18,400.00INR / Pkg of 100
Thermo Scientific SureSTART™ Vial Kit, Amber Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 8 mL; 200/Pk 0892233 Mfr # 6AK8ACT
COMPARE
Polyethylene foam/PTFE Amber 8
₹36,500.00INR / Pkg of 200
Thermo Scientific SureSTART™ Vial Kit, Amber Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 12 mL; 200/Pk 0892234 Mfr # 6AK12ACT
COMPARE
Polyethylene foam/PTFE Amber 12
₹37,700.00INR / Pkg of 200
Thermo Scientific SureSTART™ Vial Kit, Amber Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 16 mL; 200/Pk 0892236 Mfr # 6AK16ACT
COMPARE
White Polyethylene Foam/White PTFE Amber 16
₹45,300.00INR / Pkg of 200
Thermo Scientific SureSTART™ Vial Kit, Amber Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 40 mL; 100/Pk 0892239 Mfr # 6AK40ACT
COMPARE
Polyethylene foam/PTFE Amber 40
₹30,100.00INR / Pkg of 100
Thermo Scientific SureSTART™ Vial Kit, Clear Glass, Polypropylene Cap with PTFE/Silicone Septum, 20 mL; 200/Pk 0892248 Mfr # 6AK20COTAS
COMPARE
PTFE/silicone Clear 20
₹33,000.00INR / Pkg of 200
Thermo Scientific SureSTART™ Vial Kit, Amber Glass, Polypropylene Cap with Polyethylene Foam/PTFE Septum, 16 mL; 200/Pk 0892262 Mfr # 6AK16ACT
COMPARE
Polyethylene foam/PTFE Amber 16
₹37,000.00INR / Pkg of 200
Thermo Scientific SureSTART™ Vial Kit, Clear Glass, Polypropylene Cap with Silicone/PTFE Septum, 40 mL; 100/Pk   0892263 Mfr # 6AK40COTAS
COMPARE
Clear Silicone/White PTFE Clear 40
₹37,300.00INR / Pkg of 100
Thermo Scientific SureSTART™ Vial Kit, Amber Glass, Polypropylene Cap with Silicone/PTFE Septum, 40 mL; 100/Pk 0892264 Mfr # 6AK40AOTAS
COMPARE
Clear Silicone/White PTFE Amber 40
₹35,000.00INR / Pkg of 100
Showing 1- 15 of 15