Thermo Scientific Screw Cap F/Sample Vial 100/Pk 15-425 THREAD OPEN TOP W/O SEPTA
Representative image only.
Thermo Scientific Screw Cap F/Sample Vial 100/Pk 15-425 THREAD OPEN TOP W/O SEPTA
Thermo Scientific | Mfr: B7807-15| Item: 6116021
Certified performance vials are quality tested to deliver the reproducibility you need
₹4,500.00INR / Pkg of 100
₹4,500.00INR / Pkg of 100
Buy It Together
Current Item
Thermo Scientific Screw Cap F/Sample Vial 100/Pk 15-425 THREAD  OPEN TOP W/O SEPTA
Thermo Scientific Screw Cap F/Sample Vial 100/Pk 15-425 THREAD OPEN TOP W/O SEPTA
6116021
₹4,500.00INR / Pkg of 100
+ + + =
Total Price 46700.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for chromatography applications
  • Decrease your risk of failed analysis and sample reruns
  • 100% traceable through the manufacturing process
More About this Item
Meet your certification and documentation needs with the Thermo Scientific™ SureSTART™ specification certified autosampler vials. The unique threaded crown design of these glass vials ensures a secure seal with just a half-turn of the cap (caps sold separately). Precision formed neck and crimp cap profile are compatible with robotic arm autosampler trays. Vials with write-on patches have graduations at 0.5, 1.0, and 1.5 mL.
Choose from Level 2 or 3 compliance. Level 2 products have very low metal contents, extractables, and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. Level 3 products are packed in clean rooms to protect against environmental factors. They contain the lowest levels of metals, extractables, and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes.