Thermo Scientific Nalgene 5027-0909 PC Storage CryoBox, 81 x 4-5 mL Vials; 4/Pk
Representative image only.
Thermo Scientific Nalgene 5027-0909 PC Storage CryoBox, 81 x 4-5 mL Vials; 4/Pk
Thermo Scientific | Mfr: 5027-0909| Item: 0675456
₹35,000.00INR / Pkg of 4
₹35,000.00INR / Pkg of 4
Buy It Together
Current Item
Thermo Scientific Nalgene 5027-0909 PC Storage CryoBox, 81 x 4-5 mL Vials; 4/Pk
Thermo Scientific Nalgene 5027-0909 PC Storage CryoBox, 81 x 4-5 mL Vials; 4/Pk
0675456
₹35,000.00INR / Pkg of 4
+ + =
Total Price 43830.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Nalgene cryogenic vial storage boxes are ideal for mechanical freezers or liquid nitrogen. Polycarbonate (PC) boxes are vented, allowing rapid cooling of contents. Gridded insert and special lid with one-way fit make sample inventory foolproof. Boxes withstand temperatures from –196 to 121°C. Autoclavable.
1 Year Warranty
Specifications
  • Tubes Held81
  • Tube Size4.0 to 5.0 mL
  • Lowest Set Temperature (° C)-196
  • ColorBlue and Clear
  • Height (in)3 3/4
  • Height (cm)9.5
  • DescriptionPC Storage CryoBox, 81 x 4-5 mL Vials; 4/Pk