Thermo Scientific Nalgene 5026-1010 PC Storage CryoBox, 100 x 1-2 mL Vials; 10/Pk
Representative image only.
Thermo Scientific Nalgene 5026-1010 PC Storage CryoBox, 100 x 1-2 mL Vials; 10/Pk
Thermo Scientific | Mfr: 5026-1010| Item: 0675452
₹69,700.00INR / Pkg of 10
₹69,700.00INR / Pkg of 10
Buy It Together
Current Item
Thermo Scientific Nalgene 5026-1010 PC Storage CryoBox, 100 x 1-2 mL Vials; 10/Pk
Thermo Scientific Nalgene 5026-1010 PC Storage CryoBox, 100 x 1-2 mL Vials; 10/Pk
0675452
₹69,700.00INR / Pkg of 10
+ + =
Total Price 78530.00 ADD 3 ITEMS TO CART
Product Description
More About this Item
Nalgene cryogenic vial storage boxes are ideal for mechanical freezers or liquid nitrogen. Polycarbonate (PC) boxes are vented, allowing rapid cooling of contents. Gridded insert and special lid with one-way fit make sample inventory foolproof. Boxes withstand temperatures from –196 to 121°C. Autoclavable.
Specifications
  • Tubes Held100
  • Tube Size1.0 to 2.0 mL
  • Lowest Set Temperature (° C)-196
  • ColorBlue and Clear
  • Height (in)2
  • Height (cm)5.1
  • DescriptionPC Storage CryoBox, 100 x 1-2 mL Vials; 10/Pk