Thermo Scientific Nalgene 3645-2550 (R) Automatic burette adapter kit
Representative image only.
Thermo Scientific Nalgene 3645-2550 (R) Automatic burette adapter kit
Thermo Scientific | Mfr: 3645-2550| Item: 0620975
Easily adapt any conventional burette (buret)
₹12,700.00INR / Each
₹12,700.00INR / Each
Buy It Together
Current Item
Thermo Scientific Nalgene 3645-2550 (R) Automatic burette adapter kit
Thermo Scientific Nalgene 3645-2550 (R) Automatic burette adapter kit
0620975
₹12,700.00INR / Each
+ + + + =
Total Price 271800.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
1000-mL LDPE reservoir bottle with PP screw closeure, 4-ft of clear flexible tubing, PP fill tube, and burette adapter.
Key Features
  • Save time and increase convenience
  • 1 year warranty
More About this Item
This burette (buret) adapter kit allows you to convert any conventional glass or plastic, 25- or 50-mL burette into an automatic-filling, self-zeroing burette. Avoid messy spills from manual filling or funneling reagents into the top of the burette. The squeeze bottle is the reservoir for the titrant, which is delivered into the mouth of the buret through the provided clear PVC tubing.
1 Year Warranty
Specifications
  • Description(R) Automatic burette adapter kit