Thermo Scientific Hypersil 30103-014001 ODS Drop-In Guard Cartridges, 4.6/4 mm ID, 3um; 4/Pk
Representative image only.
Thermo Scientific Hypersil 30103-014001 ODS Drop-In Guard Cartridges, 4.6/4 mm ID, 3um; 4/Pk
Thermo Scientific | Mfr: 30103-014001| Item: 0188720
Protect and prolong the life of your column
₹189,000.00INR / Each
₹189,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific Hypersil 30103-014001 ODS Drop-In Guard Cartridges, 4.6/4 mm ID, 3um; 4/Pk
Thermo Scientific Hypersil 30103-014001 ODS Drop-In Guard Cartridges, 4.6/4 mm ID, 3um; 4/Pk
0188720
₹189,000.00INR / Each
+
Thermo Scientific Uniguard LID LIFT POCKET F/33850-00
0190522
₹87,500.00INR / Each
=
Total Price 276500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Easily trap contaminants such as solvent and sample debris
  • Convenient and economical
  • 1 month warranty
More About this Item
Drop-In Guard Cartridges will extend the life and performance of your column. These cartridges provide an economical and efficient way of trapping contaminants such as solvent and sample debris. Drop-in design makes for easy guard and column replacement — simply position between the injector and analytical column to remove damaging components before they reach the column. The 10-mm length offers maximum protection with minimal retention.
1 Month Warranty
Specifications
  • PhaseOctadecyl silane (C18)
  • Column Inside Diameter (ID) (mm)4
  • Column Particle Size (µm)3
  • Tubing Inside Diameter (ID) (mm)4
  • DescriptionODS Drop-In Guard Cartridges, 4.6/4 mm ID, 3um; 4/Pk
  • Warranty1 month