Thermo Scientific™ Hypersil™ ODS C18 Drop-In Guard Cartridges
Protect and prolong the life of your column
  • Easily trap contaminants such as solvent and sample debris
  • Convenient and economical
Drop-In Guard Cartridges will extend the life and performance of your column. These cartridges provide an economical and efficient way of trapping contaminants such as solvent and sample debris. Drop-in design makes for easy guard and column replacement — simply position between the injector and analytical column to remove damaging components before they reach the column. The 10-mm length offers maximum protection with minimal retention.
171,000.00 - ₹189,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Particle Size (µm) Description
Availability Pricing
Thermo Scientific Hypersil 30103-012101 ODS Drop-In Guard Cartridges, C18, 3 µm, 10 mm L x 2.1 mm ID; 4/Pk 0188718 Mfr # 30103-012101
COMPARE
2.1 3 ODS Drop-In Guard Cartridges, C18, 3 µm, 10 mm L x 2.1 mm ID; 4/Pk
₹189,000.00INR / Each
Thermo Scientific Hypersil 30103-014001 ODS Drop-In Guard Cartridges, 4.6/4 mm ID, 3um; 4/Pk 0188720 Mfr # 30103-014001
COMPARE
4 3 ODS Drop-In Guard Cartridges, 4.6/4 mm ID, 3um; 4/Pk
₹189,000.00INR / Each
Thermo Scientific Hypersil 30105-014001 ODS Drop-In Guard Cartridges, C18, 5 µm, 10 mm L x 4.0 mm ID; 4/Pk 0188744 Mfr # 30105-014001
COMPARE
4 5 ODS Drop-In Guard Cartridges, C18, 5 µm, 10 mm L x 4.0 mm ID; 4/Pk
₹171,000.00INR / Each