Thermo Scientific Autosampler Vial Septa, 13 mm, PTFE, White Virgin PTFE Pre-slit Septum; 100/Pk
Representative image only.
Thermo Scientific Autosampler Vial Septa, 13 mm, PTFE, White Virgin PTFE Pre-slit Septum; 100/Pk
Thermo Scientific | Mfr: C4015-10| Item: 9881421
Keep your samples safe with these vial closures
₹17,100.00INR / Pkg
₹17,100.00INR / Pkg
Buy It Together
Current Item
Thermo Scientific Autosampler Vial Septa, 13 mm, PTFE, White Virgin PTFE Pre-slit Septum; 100/Pk
Thermo Scientific Autosampler Vial Septa, 13 mm, PTFE, White Virgin PTFE Pre-slit Septum; 100/Pk
9881421
₹17,100.00INR / Pkg
+ + + + + =
Total Price 102900.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Certified performance caps are quality tested to deliver the reproducibility you need
  • Decrease your risk of failed analysis and sample reruns
  • Easily secure vial contents with screw, snap, or crimp caps
  • For use with 13-425 screw caps
More About this Item
Meet your certification and documentation needs with the Thermo Scientific™ SureSTART™ specification certified vial caps. Level 1 products are cost optimized, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. Level 2 products have very low metal contents, extractables, and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. Level 3 products are packed in clean rooms to protect against environmental factors. They contain the lowest levels of metals, extractables, and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes.
Caps are available in different closure types, colors, and septum materials to suit your application. AVCS™ screw caps incorporate Advanced Vial Closure System technology to ensure optimal seal compression when closing a vial. Snap caps can be snapped on and off by hand which eliminates the need for metal crimping and decapping tools. Crimp caps crimps tightly around the neck for a tight seal.
Specifications
  • Cap TypeNone
  • Cap Diameter (mm)13
  • Cap MaterialPTFE
  • Cap colorWhite
  • Septum TypeWhite Virgin PTFE
  • DescriptionAutosampler Vial Septa, 13 mm, PTFE, White Virgin PTFE Pre-slit Septum; 100/Pk