Testo 0563 1052 Thermometer, T-Handle, Kit (Includes 3 Probes)
Representative image only.
Testo 0563 1052 Thermometer, T-Handle, Kit (Includes 3 Probes)
Testo | Mfr: 0563 1052| Item: 9260014
Three easy-to-interchange probes for various food applications
₹126,000.00INR / Each
for an additional
₹30,000.00INR / Each
for an additional
₹42,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹126,000.00INR / Each
Buy It Together
Current Item
Testo 0563 1052 Thermometer, T-Handle, Kit (Includes 3 Probes)
Testo 0563 1052 Thermometer, T-Handle, Kit (Includes 3 Probes)
9260014
₹126,000.00INR / Each
+
InnoCal NIST-Traceable Calibration; Digital Indicator
1710161
₹30,000.00INR / Each
=
Total Price 156000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Ergonomic design provides comfort during repetitive measurements
  • 1 year warranty
More About this Item
These food thermometers were developed specifically for food professionals. The robust, IP65-rated body is water- and dust-resistant for high liquid or moisture environments. User-selectable high and low alarm limits provide an audible and visual alarm when critical thresholds are exceeded. The backlit display allows convenient viewing in low-lit areas.
Model 92600-14 offers the complete measurement package with three interchangeable probes for a variety of food temperature measurement applications.
1 Year WarrantyIP 65
Specifications
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° F)-58
  • Max Temperature (° F)525
  • Min Temperature (° C)-50
  • Max Temperature (° C)275
  • Resolution0.1°
  • IP RatingIP65
  • Display TypeLCD
  • Height (in)5 3/4
  • Width (in)7 1/2
  • Battery Life80 hours
  • Battery4 1.5V cell (included)
  • DescriptionThermometer, T-Handle, Kit (Includes 3 Probes)