InnoCal NIST-Traceable Calibration; Digital Indicator
Representative image only.
InnoCal NIST-Traceable Calibration; Digital Indicator
InnoCal | Item: 1710161
NIST-traceable calibrations for temperature meters
₹30,000.00INR / Each
₹30,000.00INR / Each
Buy It Together
Current Item
InnoCal NIST-Traceable Calibration; Digital Indicator
InnoCal NIST-Traceable Calibration; Digital Indicator
1710161
₹30,000.00INR / Each
+ + + + + + + + + =
Total Price 573500.00 ADD 10 ITEMS TO CART
Product Description
Includes
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated.
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Certificate lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • 1 year warranty
More About this Item
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.
Calibrations on handheld digital indicators are performed using direct comparison in high-stability liquid and dry baths using high-precision PRT’s and SPRT’s and displays. Measurements can be performed at four points between -80 deg C and 1300 deg C. Points are generally selected at standard ITS-90 points across the range of the indicator but customer-specified points can be accommodated.
NIST Traceable Calibration1 Year Warranty
Specifications
  • Calibration TypeTemperature
  • Instrument Only CalibrationNo
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points4 points
  • Calibration Test PointsITS-90 points across the range unless specfied
  • Calibration Test Range-196 to 1300 C
  • DescriptionNIST-Traceable Calibration; Digital Indicator