Testo Frozen Food Thermometer with Interchangeable Probes
Three easy-to-interchange probes for various food applications
  • Ergonomic design provides comfort during repetitive measurements
These food thermometers were developed specifically for food professionals. The robust, IP65-rated body is water- and dust-resistant for high liquid or moisture environments. User-selectable high and low alarm limits provide an audible and visual alarm when critical thresholds are exceeded. The backlit display allows convenient viewing in low-lit areas.
42,100.00 - ₹86,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Number Of Probe Channels
Availability Pricing
Testo 0563 1051 T-Handle Digital Thermometer; -50-275C/-58-525F WW-92600-12 Mfr # 0563 1051
COMPARE
525 Digital Indicator 1
₹42,100.00INR / Each
Testo 0563 1052 Thermometer, T-Handle, Kit (Includes 3 Probes) WW-92600-14 Mfr # 0563 1052
COMPARE
525 Digital Indicator 1
₹86,500.00INR / Each