Sherwood Scientific 50035006 Fluid Bed Dryer Tub and Assembly, Non-Sealed, Glass, 2-Liter
Representative image only.
Sherwood Scientific 50035006 Fluid Bed Dryer Tub and Assembly, Non-Sealed, Glass, 2-Liter
Sherwood Scientific | Mfr: 50035006| Item: 0265852
Quickly and evenly dry particles as small as 5 μm
Call For Price
Call For Price
Buy It Together
Current Item
Sherwood Scientific 50035006 Fluid Bed Dryer Tub and Assembly, Non-Sealed, Glass, 2-Liter
Sherwood Scientific 50035006 Fluid Bed Dryer Tub and Assembly, Non-Sealed, Glass, 2-Liter
0265852
Call For Price
+ + + + + =
Total Price 709000.00 ADD 6 ITEMS TO CART
Product Description
Key Features
  • Faster and gentler than conventional drying cabinets—ideal for food products, polymers, soils, chemicals, pharmaceuticals, minerals, and waste materials
  • Variety of accessories provide easy customization for your drying applications
More About this Item
Non-Sealed Tub Assemblies are available in glass or stainless steel and require a filter bag, sold separately below.
Model 02658-02 includes a port to add extra sample and a port for a temperature/humidity probe (sold separately).
Order fluid bed dryer, filter bags, and optional accessories separately for a complete fluid bed dryer system.
Glass tubs are useful for observation of applications and determination of the optimum fluidizing velocity. The 2- and 5-L units handle samples up to 11 lb (5.0 kg). Required filter bags sold separately.
†Base (02658-40/45) must be ordered separately.
Specifications
  • Materials of ConstructionGlass
  • DescriptionFluid Bed Dryer Tub and Assembly, Non-Sealed, Glass, 2-Liter