Sherwood Scientific 50186500 Temperature/Humidity Probe for Fluid Bed Dryers
Representative image only.
Sherwood Scientific 50186500 Temperature/Humidity Probe for Fluid Bed Dryers
Sherwood Scientific | Mfr: 50186500| Item: 0265811
Optimize performance of your Sherwood Scientific fluid bed dryer
₹709,000.00INR / Each
₹709,000.00INR / Each
Buy It Together
Current Item
Sherwood Scientific 50186500 Temperature/Humidity Probe for Fluid Bed Dryers
Sherwood Scientific 50186500 Temperature/Humidity Probe for Fluid Bed Dryers
0265811
₹709,000.00INR / Each
+ + + + + =
Total Price 709000.00 ADD 6 ITEMS TO CART
Product Description
Key Features
  • Keep your fluid bed dryer running properly with these accessories
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Sherwood Scientific fluid bed dryer.
The Temperature/Humidity probe allows you to control the drying by ending a program step at a specific temperature or humidity.
1 Year Warranty
Specifications
  • DescriptionTemperature/Humidity Probe for Fluid Bed Dryers