Sherwood Scientific 50035402 Filter Bag for Non-Sealed Tubs, Large, Terylene
Representative image only.
Sherwood Scientific 50035402 Filter Bag for Non-Sealed Tubs, Large, Terylene
Sherwood Scientific | Mfr: 50035402| Item: 0265884
Safely retains stray particles
Call For Price
Call For Price
Buy It Together
Current Item
Sherwood Scientific 50035402 Filter bag for non-sealed tubs, Large, terylene
Sherwood Scientific 50035402 Filter bag for non-sealed tubs, Large, terylene
0265884
Call For Price
+ + + =
Total Price 709000.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Allows passage of exit gases
More About this Item
Bags retain stray particles of the sample being fluidized in the tub while allowing the passage of exit gases. Use one large bag for a single two- or five-liter tub assembly; use four small bags for the multi-tub assemblies.
Specifications
  • DescriptionFilter bag for non-sealed tubs, Large, terylene