Sellstrom 39110 Acetate Face Shield with Headgear, Clear Shield
Representative image only.
Sellstrom 39110 Acetate Face Shield with Headgear, Clear Shield
Sellstrom | Mfr: S39110| Item: 8647816
The chemical protection of acetate at an economical price
₹12,700.00INR / Each
₹12,700.00INR / Each
Buy It Together
Current Item
Sellstrom 39110 Acetate Face Shield with Headgear, Clear Shield
Sellstrom 39110 Acetate Face Shield with Headgear, Clear Shield
8647816
₹12,700.00INR / Each
+
Pyrex Vista 70000-250 Griffin Style Beakers, 250 mL 12/pk
3453064
₹10,900.00INR / Pkg of 12
+ =
Total Price 27700.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Ratcheting head gear allows for fast and easy adjustment. Face shield curves naturally to fit facial contours ensuring reliable protection from splash, particles, and flying debris. Acetate window is chemical splash, and impact resistant. Window easily attaches with keyhole design and locks in the up or down position depending on need. Nonglare finish allows for high clarity.
Compliance: ANSI Z87.1-2003, CUL, CAN/CSA-Z94.3 Standards
1 Year Warranty
Specifications
  • Frame ColorBlue
  • Lens ColorClear
  • Lens CoatingNone
  • ComplianceANSI Z87.1-2003; cUL, CAN/CSA-Z94.3
  • DescriptionAcetate Face Shield with Headgear, Clear Shield