PYREX® VISTA™ Griffin Low-Form Glass Beakers
Get a reliable measurement at an affordable price
  • No mystery liquids—large marking spot for easy identification
Economical low-form beakers feature a pouring spout and a large marking spot with a ±5% graduations. Reference: ASTM E-960.
9,000.00 - ₹15,600.00INR / Pkg of 12
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Pyrex Vista 70000-10 Griffin Style Beakers, 10 mL, 12/pk 3453052 Mfr # 70000-10
COMPARE
10 0 Glass
₹13,800.00INR / Pkg of 12
Pyrex Vista 70000-20 Griffin Style Beakers, 20 mL 12/pk 3453054 Mfr # 70000-20
COMPARE
20 5 Glass
₹12,200.00INR / Pkg of 12
Pyrex Vista 70000-30 Griffin Style Beakers, 30 mL 12/pk 3453056 Mfr # 70000-30
COMPARE
30 10 Glass
₹11,300.00INR / Pkg of 12
Pyrex Vista 70000-50 Griffin Style Beakers, 50 mL 12/pk 3453058 Mfr # 70000-50
COMPARE
50 10 Glass
₹11,300.00INR / Pkg of 12
Pyrex Vista 70000-100 Griffin Style Beakers, 100 mL 12/pk 3453060 Mfr # 70000-100
COMPARE
100 10 Glass
₹11,700.00INR / Pkg of 12
Pyrex Vista 70000-150 Griffin Style Beakers, 150 mL 12/pk 3453062 Mfr # 70000-150
COMPARE
150 10 Glass
₹11,700.00INR / Pkg of 12
Pyrex Vista 70000-250 Griffin Style Beakers, 250 mL 12/pk 3453064 Mfr # 70000-250
COMPARE
250 25 Glass
₹10,900.00INR / Pkg of 12
Pyrex Vista 70000-400 Griffin Style Beakers, 400 mL 12/pk 3453066 Mfr # 70000-400
COMPARE
400 25 Glass
₹13,800.00INR / Pkg of 12
Pyrex Vista 70000-600 Griffin Style Beakers, 600 mL 6/pk 3453068 Mfr # 70000-600
COMPARE
600 50 Glass
₹9,000.00INR / Pkg of 6
Pyrex Vista 70000-1L Griffin Style Beakers, 1000 mL 6/pk 3453070 Mfr # 70000-1L
COMPARE
1000 50 Glass
₹15,600.00INR / Pkg of 6
Showing 1- 10 of 10