Sellstrom Acetate Face Shield
The chemical protection of acetate at an economical price
Ratcheting head gear allows for fast and easy adjustment. Face shield curves naturally to fit facial contours ensuring reliable protection from splash, particles, and flying debris. Acetate window is chemical splash, and impact resistant. Window easily attaches with keyhole design and locks in the up or down position depending on need. Nonglare finish allows for high clarity.
Compliance: ANSI Z87.1-2003, CUL, CAN/CSA-Z94.3 Standards
12,700.00 - ₹14,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Frame Color Lens Coating Lens Color
Availability Pricing
Sellstrom 39110 Acetate Face Shield with Headgear, Clear Shield 8647816 Mfr # S39110
COMPARE
Blue None Clear
₹12,700.00INR / Each
Sellstrom 39140 Acetate Face Shield with Headgear, Clear Anti-Fog Shield 8647817 Mfr # S39140
COMPARE
Blue Anti-fog Clear
₹14,600.00INR / Each