Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 3/4" Mini Tri-Clamp x 3/4" ID; 1/Each
Representative image only.
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 3/4" Mini Tri-Clamp x 3/4" ID; 1/Each
SANI-TECH | Mfr: TAD075-12H| Item: 5013469
Effortlessly connect flexible tubing to sanitary ferrules
₹15,000.00INR / Each
₹15,000.00INR / Each
Buy It Together
Current Item
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 3/4" Mini Tri-Clamp x 3/4" ID; 1/Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 3/4" Mini Tri-Clamp x 3/4" ID; 1/Each
5013469
₹15,000.00INR / Each
+ + + + =
Total Price 239700.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Crafted from high-quality polypropylene to ensure ultra-pure water and chemical transfer
  • Resistant to corrosion, pitting, and scaling while offering excellent chemical resistance
  • Meets USP Class VI and 21 CFR standards for ultimate safety and compliance
More About this Item
Sani-Tech® sanitary adapters provide the ideal solution for integrating different connection technologies into your system. Designed for industries requiring the highest purity, these adapters prevent biofilm growth more effectively than stainless steel alternatives. Operating at temperatures up to 125°F, they deliver unmatched durability and reliability for pharmaceutical, food & beverage, dairy, and ultra-pure water applications.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolypropylene
  • Connection TypeSanitary x Hose barb
  • Barb Size / Tubing Id (Port 1) (in)3/4
  • Sanitary Clamp Size (Port 1) (InchShort)3/4
  • Flow PatternStraight
  • Max Temperature (° F)125
  • Max Temperature (° C)51.67
  • DescriptionSanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 3/4" Mini Tri-Clamp x 3/4" ID; 1/Each
  • WarrantyManufacturer defect