Sani-Tech® Hose Barb Adapters, Polypropylene
Effortlessly connect flexible tubing to sanitary ferrules
  • Crafted from high-quality polypropylene to ensure ultra-pure water and chemical transfer
  • Resistant to corrosion, pitting, and scaling while offering excellent chemical resistance
  • Meets USP Class VI and 21 CFR standards for ultimate safety and compliance
Sani-Tech® sanitary adapters provide the ideal solution for integrating different connection technologies into your system. Designed for industries requiring the highest purity, these adapters prevent biofilm growth more effectively than stainless steel alternatives. Operating at temperatures up to 125°F, they deliver unmatched durability and reliability for pharmaceutical, food & beverage, dairy, and ultra-pure water applications.
15,000.00 - ₹31,400.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Sanitary Clamp Size (Port 1) (InchShort)
Availability Pricing
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 3/4" Mini Tri-Clamp x 1/8" ID; 1/Each 5013465 Mfr # TAD075-02H
COMPARE
1/8 3/4
₹15,000.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 3/4" Mini Tri-Clamp x 1/4" ID; 1/Each 5013466 Mfr # TAD075-04H
COMPARE
1/4 3/4
₹15,500.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 3/4" Mini Tri-Clamp x 3/8" ID; 1/Each 5013467 Mfr # TAD075-06H
COMPARE
3/8 3/4
₹15,000.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 3/4" Mini Tri-Clamp x 1/2" ID; 1/Each 5013468 Mfr # TAD075-08H
COMPARE
1/2 3/4
₹15,000.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 3/4" Mini Tri-Clamp x 3/4" ID; 1/Each 5013469 Mfr # TAD075-12H
COMPARE
3/4 3/4
₹15,000.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 1" Ladish Tri-Clamp x 1/8" ID; 1/Each 5013470 Mfr # TAD100L-02H
COMPARE
1/8 1
₹31,400.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 1" Ladish Tri-Clamp x 1/4" ID; 1/Each 5013471 Mfr # TAD100L-04H
COMPARE
1/4 1
₹29,900.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 1" Ladish Tri-Clamp x 3/8" ID; 1/Each 5013472 Mfr # TAD100L-06H
COMPARE
3/8 1
₹31,400.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 1" Ladish Tri-Clamp x 1/2" ID; 1/Each 5013473 Mfr # TAD100L-08H
COMPARE
1/2 1
₹29,900.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 1" Ladish Tri-Clamp x 3/4" ID; 1/Each 5013474 Mfr # TAD100L-12H
COMPARE
3/4 1
₹29,900.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 1" Ladish Tri-Clamp x 1" ID; 1/Each 5013475 Mfr # TAD100L-16H
COMPARE
1 1
₹31,400.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 1-1/2" Tri-Clamp x 1/4" ID; 1/Each 5013476 Mfr # TAD150-04H
COMPARE
1/4 1-1/2
₹29,900.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 1-1/2" Tri-Clamp x 3/8" ID; 1/Each 5013477 Mfr # TAD150-06H
COMPARE
3/8 1-1/2
₹29,900.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 1-1/2" Tri-Clamp x 1/2" ID; 1/Each 5013478 Mfr # TAD150-08H
COMPARE
1/2 1-1/2
₹29,900.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 1-1/2" Tri-Clamp x 1" ID; 1/Each 5013479 Mfr # TAD150-16H
COMPARE
1 1-1/2
₹31,000.00INR / Each
Sani-Tech® Sanitary to Hose Barb Adapter, Straight Adapter, Polypropylene, 1-1/2" Tri-Clamp x 1-1/2" ID; 1/Each 5013480 Mfr # TAD150-24H
COMPARE
1 1/2 1-1/2
₹29,900.00INR / Each
Showing 1- 16 of 16