QLA Straight 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 5.9"L, 1/8" OD; SS Luer Lock
Representative image only.
QLA Straight 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 5.9"L, 1/8" OD; SS Luer Lock
| Mfr: CA8475-VKS| Item: 4130205
Achieve consistent sampling with QLA cannulas
₹32,300.00INR / Each
₹32,300.00INR / Each
Buy It Together
Current Item
QLA Straight 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 5.9"L, 1/8" OD; SS Luer Lock
QLA Straight 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 5.9"L, 1/8" OD; SS Luer Lock
4130205
₹32,300.00INR / Each
+ =
Total Price 49600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency
  • 5.9"L straight SS cannula
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannulas and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
Parts are made of 316 stainless steel that will not absorb or interfere with active ingredients.
.
Specifications
  • Material316 stainless steel
  • Outside Diameter (OD) (mm)3.2
  • Length (")5 29/32
  • DescriptionStraight 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 5.9"L, 1/8" OD; SS Luer Lock