QLA Adjustable Cannula Stopper with PTFE Sleeve Compatible with Agilent Dissolution Apparatus; 1/8" (3.2 mm) OD
Representative image only.
QLA Adjustable Cannula Stopper with PTFE Sleeve Compatible with Agilent Dissolution Apparatus; 1/8" (3.2 mm) OD
| Mfr: CA8STP-VK| Item: 4130209
QLA accessories help provide more precise and consistent dissolution results
₹16,500.00INR / Each
₹16,500.00INR / Each
Buy It Together
Current Item
QLA Adjustable Cannula Stopper with PTFE Sleeve Compatible with Agilent Dissolution Apparatus; 1/8" (3.2 mm) OD
QLA Adjustable Cannula Stopper with PTFE Sleeve Compatible with Agilent Dissolution Apparatus; 1/8" (3.2 mm) OD
4130209
₹16,500.00INR / Each
+ =
Total Price 33800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • For Agilent
  • Comes with PTFE sleeve
More About this Item
Use stopper with cannula to ensure the sample is always taken from the same depth.
.
Specifications
  • Compatible WithAgilent apparatus
  • DescriptionAdjustable Cannula Stopper with PTFE Sleeve Compatible with Agilent Dissolution Apparatus; 1/8" (3.2 mm) OD