QLA Adjustable Cannula Stopper Compatible with Agilent 708/709-DS Dissolution Apparatus; 1/8" (3.2 mm) OD
Representative image only.
QLA Adjustable Cannula Stopper Compatible with Agilent 708/709-DS Dissolution Apparatus; 1/8" (3.2 mm) OD
| Mfr: CANSTP125-70XDS| Item: 4130274
QLA accessories help provide more precise and consistent dissolution results
₹17,300.00INR / Each
₹17,300.00INR / Each
Product Description
Key Features
  • For Agilent 708/709-DS
  • 1/8" (3.2 mm) OD
More About this Item
Use stopper with cannula to ensure the sample is always taken from the same depth.
.
Specifications
  • Compatible WithAgilent 708/709-DS
  • DescriptionAdjustable Cannula Stopper Compatible with Agilent 708/709-DS Dissolution Apparatus; 1/8" (3.2 mm) OD