QLA Dual DV Series Sampling Probe Compatible with Agilent and VanKel EaseAlign Baths; for 900 mL Vessel
Representative image only.
QLA Dual DV Series Sampling Probe Compatible with Agilent and VanKel EaseAlign Baths; for 900 mL Vessel
| Mfr: CAN900-DVV| Item: 4130260
Achieve consistent sampling with QLA cannulas
₹57,100.00INR / Each
₹57,100.00INR / Each
Buy It Together
Current Item
QLA Dual DV Series Sampling Probe Compatible with Agilent and VanKel EaseAlign Baths; for 900 mL Vessel
QLA Dual DV Series Sampling Probe Compatible with Agilent and VanKel EaseAlign Baths; for 900 mL Vessel
4130260
₹57,100.00INR / Each
+ =
Total Price 74100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency
  • Features a return port
  • Use with 900 mL vessel
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannulas and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
Dual DV series probe features a return port for dissolution protocol that require samples to be returned to the vessel. Since both the collection and return ports are in the same unit, only one port in the vessel cover is used. The probe remains in the vessel throughout the dissolution test so you can sample from the same position every time. A twist-lock housing accepts a standard full flow filter.
.
Specifications
  • Material316 SS cannula
  • DescriptionDual DV Series Sampling Probe Compatible with Agilent and VanKel EaseAlign Baths; for 900 mL Vessel