QLA Adjustable Cannula Stopper for VanKel/Varian; 0.082" OD

Representative image only.

QLA Adjustable Cannula Stopper for VanKel/Varian; 0.082" OD

| Mfr: CANSTP-VK| Item: 4130282

QLA accessories help provide more precise and consistent dissolution results

₹18,500.00INR / Each

₹18,500.00INR / Each
Product Description

Key Features

  • for VanKel/Varian
  • 0 082" OD

More About this Item

Use stopper with cannula to ensure the sample is always taken from the same depth.

.

Specifications
  • Compatible WithVanKel/Varian apparatus
  • DescriptionAdjustable Cannula Stopper for VanKel/Varian; 0.082" OD
Reviews
Q&A