QLA Bent PEEK Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, for 900 mL vessel, 4.75"L, 1/8" OD; PEEK Luer Lock
Representative image only.
QLA Bent PEEK Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, for 900 mL vessel, 4.75"L, 1/8" OD; PEEK Luer Lock
| Mfr: CA8475-PV| Item: 4130203
Achieve consistent sampling with QLA cannulas
₹33,400.00INR / Each
₹33,400.00INR / Each
Buy It Together
Current Item
QLA Bent PEEK Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, for 900 mL vessel, 4.75"L, 1/8" OD; PEEK Luer Lock
QLA Bent PEEK Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, for 900 mL vessel, 4.75"L, 1/8" OD; PEEK Luer Lock
4130203
₹33,400.00INR / Each
+ =
Total Price 50700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency
  • 4.75"L for 900 mL vessel
  • PEEK construction
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannulas and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
Parts are made of PEEK so they won't absorb or affect ingredients.
.
Specifications
  • MaterialPEEK
  • Outside Diameter (OD) (mm)3.2
  • Length (")4 3/4
  • DescriptionBent PEEK Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, for 900 mL vessel, 4.75"L, 1/8" OD; PEEK Luer Lock